3-(Dodecanoylamino)benzoic acid

Supplier: Matrix Scientific

061373-500MG
101921-840EA 189.75 USD
101921-840
3-(Dodecanoylamino)benzoic acid
3-(Dodecanoylamino)benzoic acid

Hazard: Irritant

Formula: C₁₉H₂₉NO₃
MDL Number: MFCD00183873
CAS Number: 79564-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR