4-Acetylphenyl-4-bromobenzoate

Supplier: Matrix Scientific

061371-500MG
101921-838EA 146.15 USD
101921-838
4-Acetylphenyl-4-bromobenzoate
4-Acetylphenyl-4-bromobenzoate

Hazard: Irritant

Formula: C₁₅H₁₁BrO₃
MDL Number: MFCD00432481
CAS Number: 305857-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR