2-Bromo-N-[2-chloro-5-(trifluoromethyl)phenyl]acetamide

Supplier: Matrix Scientific

061367-500MG
101921-830EA 243.08 USD
101921-830
2-Bromo-N-[2-chloro-5-(trifluoromethyl)phenyl]acetamide
2-Bromo-N-[2-chloro-5-(trifluoromethyl)phenyl]acetamide

Hazard: Irritant

Formula: C₉H₆BrClF₃NO
MW: 316.51 g/mol
MDL Number: MFCD00506399
CAS Number: 2003-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR