2-(3-Chloro-2-methylphenyl)-1,3-dioxoisoindoline-5-carboxylic acid

Supplier: Matrix Scientific

061366-500MG
101921-828EA 243.08 USD
101921-828
2-(3-Chloro-2-methylphenyl)-1,3-dioxoisoindoline-5-carboxylic acid
2-(3-Chloro-2-methylphenyl)-1,3-dioxoisoindoline-5-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₀ClNO₄
MDL Number: MFCD02905231
CAS Number: 356576-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR