2-(1,3-Dioxooctahydro-2H-4,7-methanoisoindol-2-yl)-3-phenylpropanoic acid

Supplier: Matrix Scientific

061365-500MG
101921-826EA 364.62 USD
101921-826
2-(1,3-Dioxooctahydro-2H-4,7-methanoisoindol-2-yl)-3-phenylpropanoic acid
2-(1,3-Dioxooctahydro-2H-4,7-methanoisoindol-2-yl)-3-phenylpropanoic acid

Hazard: Irritant

Formula: C₁₈H₁₉NO₄
MDL Number: MFCD12197718
CAS Number: 1008577-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR