2-(4-Nitrophenyl)-1,3-dioxoisoindoline-5-carboxylic acid
Supplier: Matrix Scientific
061364-500MG
101921-824EA
243.08
USD
101921-824
2-(4-Nitrophenyl)-1,3-dioxoisoindoline-5-carboxylic acid
2-(4-Nitrophenyl)-1,3-dioxoisoindoline-5-carboxylic acid
Hazard: Irritant
Formula:
C₁₅H₈N₂O₆ |
MDL Number:
MFCD00187716 CAS Number: 110768-20-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...