2-(4-Nitro-1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)benzoic acid

Supplier: Matrix Scientific

061363-500MG
101921-822EA 243.08 USD
101921-822
2-(4-Nitro-1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)benzoic acid
2-(4-Nitro-1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)benzoic acid

Hazard: Irritant

Formula: C₁₅H₈N₂O₆
MDL Number: MFCD00391879
CAS Number: 100914-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR