5,6-Dibromohexahydro-2-benzofuran-1,3-dione

Supplier: Matrix Scientific

061362-500MG
101921-820EA 243.08 USD
101921-820
5,6-Dibromohexahydro-2-benzofuran-1,3-dione
5,6-Dibromohexahydro-2-benzofuran-1,3-dione

Hazard: Irritant

Formula: C₈H₈Br₂O₃
MDL Number: MFCD08445355
CAS Number: 23893-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR