5-(6-Chloropyridazin-3-yl)-2-ethyl-N-methylbenzenesulfonamide

Supplier: Matrix Scientific

061361-500MG
101921-818EA 484.62 USD
101921-818
5-(6-Chloropyridazin-3-yl)-2-ethyl-N-methylbenzenesulfonamide
5-(6-Chloropyridazin-3-yl)-2-ethyl-N-methylbenzenesulfonamide

Hazard: Irritant

Formula: C₁₃H₁₄ClN₃O₂S
MDL Number: MFCD12197717
CAS Number: 1119425-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR