2-(3-Carboxyphenyl)-1,3-dioxoisoindoline-5-carboxylic acid

Supplier: Matrix Scientific

061360-500MG
101921-816EA 364.62 USD
101921-816
2-(3-Carboxyphenyl)-1,3-dioxoisoindoline-5-carboxylic acid
2-(3-Carboxyphenyl)-1,3-dioxoisoindoline-5-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₉NO₆
MDL Number: MFCD00391887
CAS Number: 38250-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR