2-(4-Acetylphenyl)-1,3-dioxoisoindoline-5-carboxylic acid

Supplier: Matrix Scientific

061358-500MG
101921-812EA 243.08 USD
101921-812
2-(4-Acetylphenyl)-1,3-dioxoisoindoline-5-carboxylic acid
2-(4-Acetylphenyl)-1,3-dioxoisoindoline-5-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₁₁NO₅
MDL Number: MFCD00391889
CAS Number: 300405-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR