2-Chloro-5-(6-oxo-1,6-dihydropyridazin-3-yl)benzenesulfonyl chloride

Supplier: Matrix Scientific

061356-500MG
101921-808EA 484.62 USD
101921-808
2-Chloro-5-(6-oxo-1,6-dihydropyridazin-3-yl)benzenesulfonyl chloride
2-Chloro-5-(6-oxo-1,6-dihydropyridazin-3-yl)benzenesulfonyl chloride

Hazard: Irritant

Formula: C₁₀H₆Cl₂N₂O₃S
MDL Number: MFCD12197716
CAS Number: 1160264-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR