2-(4-Chlorophenyl)-1-methyl-2-oxoethyl-4-aminobenzoate

Supplier: Matrix Scientific

061355-500MG
101921-806EA 364.62 USD
101921-806
2-(4-Chlorophenyl)-1-methyl-2-oxoethyl-4-aminobenzoate
2-(4-Chlorophenyl)-1-methyl-2-oxoethyl-4-aminobenzoate

Hazard: Irritant

Formula: C₁₆H₁₄ClNO₃
MDL Number: MFCD12197715
CAS Number: 1160264-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR