2-(1,3-Dioxo-1,3,3a,4,7,7a-hexahydro-2H-isoindol-2-yl)-3-phenylpropanoic acid

Supplier: Matrix Scientific

061353-500MG
101921-802EA 364.62 USD
101921-802
2-(1,3-Dioxo-1,3,3a,4,7,7a-hexahydro-2H-isoindol-2-yl)-3-phenylpropanoic acid
2-(1,3-Dioxo-1,3,3a,4,7,7a-hexahydro-2H-isoindol-2-yl)-3-phenylpropanoic acid

Hazard: Irritant

Formula: C₁₇H₁₇NO₄
MDL Number: MFCD09735585
CAS Number: 1104074-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR