(1,3-Dioxo-5-phenyloctahydro-2H-4,7-methanoisoindol-2-yl)acetic acid

Supplier: Matrix Scientific

061352-500MG
101921-800EA 364.62 USD
101921-800
(1,3-Dioxo-5-phenyloctahydro-2H-4,7-methanoisoindol-2-yl)acetic acid
(1,3-Dioxo-5-phenyloctahydro-2H-4,7-methanoisoindol-2-yl)acetic acid

Hazard: Irritant

Formula: C₁₇H₁₇NO₄
MDL Number: MFCD06128095
CAS Number: 1218360-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR