5-(6-Chloropyridazin-3-yl)-2-ethylbenzenesulfonamide

Supplier: Matrix Scientific

061350-500MG
101921-796EA 364.62 USD
101921-796
5-(6-Chloropyridazin-3-yl)-2-ethylbenzenesulfonamide
5-(6-Chloropyridazin-3-yl)-2-ethylbenzenesulfonamide

Hazard: Irritant

Formula: C₁₂H₁₂ClN₃O₂S
MDL Number: MFCD12197713
CAS Number: 1119441-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR