2-(2,3-Dimethylphenyl)-1,3-dioxoisoindoline-5-carboxylic acid

Supplier: Matrix Scientific

061348-500MG
101921-792EA 183.08 USD
101921-792
2-(2,3-Dimethylphenyl)-1,3-dioxoisoindoline-5-carboxylic acid
2-(2,3-Dimethylphenyl)-1,3-dioxoisoindoline-5-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₁₃NO₄
MDL Number: MFCD00404566
CAS Number: 294667-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR