1-[(4-Nitrobenzoyl)amino]-5-oxopyrrolidine-3-carboxylic acid

Supplier: Matrix Scientific

061345-500MG
101921-786EA 364.62 USD
101921-786
1-[(4-Nitrobenzoyl)amino]-5-oxopyrrolidine-3-carboxylic acid
1-[(4-Nitrobenzoyl)amino]-5-oxopyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₁N₃O₆
MDL Number: MFCD06128439
CAS Number: 894622-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR