5-Oxo-1-[4-(propoxycarbonyl)phenyl]pyrrolidine-3-carboxylic acid

Supplier: Matrix Scientific

061343-500MG
101921-782EA 364.62 USD
101921-782
5-Oxo-1-[4-(propoxycarbonyl)phenyl]pyrrolidine-3-carboxylic acid
5-Oxo-1-[4-(propoxycarbonyl)phenyl]pyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₇NO₅
MDL Number: MFCD05096181
CAS Number: 899762-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR