5-Amino-2-(4-aminophenyl)-1H-isoindole-1,3(2H)-dione

Supplier: Matrix Scientific

061297-500MG
101921-694EA 484.62 USD
101921-694
5-Amino-2-(4-aminophenyl)-1H-isoindole-1,3(2H)-dione
5-Amino-2-(4-aminophenyl)-1H-isoindole-1,3(2H)-dione

Hazard: Irritant

Formula: C₁₄H₁₁N₃O₂
MDL Number: MFCD00430450
CAS Number: 13406-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR