7-(Isopropoxycarbonyl)tricyclo[3.2.2.0~2,4~]non-8-ene-6-carboxylic acid

Supplier: Matrix Scientific

061294-500MG
101921-688EA 484.62 USD
101921-688
7-(Isopropoxycarbonyl)tricyclo[3.2.2.0~2,4~]non-8-ene-6-carboxylic acid
7-(Isopropoxycarbonyl)tricyclo[3.2.2.0~2,4~]non-8-ene-6-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₈O₄
MW: 250.3 g/mol
MDL Number: MFCD12197706
CAS Number: 1218447-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR