2-Bromo-N-(1-phenylethyl)acetamide

Supplier: Matrix Scientific

061282-500MG
101921-664EA 110.77 USD
101921-664
2-Bromo-N-(1-phenylethyl)acetamide
2-Bromo-N-(1-phenylethyl)acetamide

Hazard: Irritant

Formula: C₁₀H₁₂BrNO
Melting Pt: 82…83 °C
MDL Number: MFCD05624926
CAS Number: 70110-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR