2-Oxo-4-phenyl-1,2,3,4-tetrahydropyrimidine-5-carbohydrazide

Supplier: Matrix Scientific

061276-500MG
101921-652EA 484.62 USD
101921-652
2-Oxo-4-phenyl-1,2,3,4-tetrahydropyrimidine-5-carbohydrazide
2-Oxo-4-phenyl-1,2,3,4-tetrahydropyrimidine-5-carbohydrazide

Hazard: Irritant

Formula: C₁₁H₁₂N₄O₂
MDL Number: MFCD12197705
CAS Number: 1160264-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR