4-Bromo-2,6-diethylaniline

Supplier: Matrix Scientific

061273-500MG
101921-646EA 484.62 USD
101921-646
4-Bromo-2,6-diethylaniline
4-Bromo-2,6-diethylaniline

Hazard: Irritant

Formula: C₁₀H₁₄BrN
MDL Number: MFCD00059266
CAS Number: 56746-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR