4-Amino-N-(2-methylphenyl)benzamide

Supplier: Matrix Scientific

061268-500MG
101921-636EA 146.15 USD
101921-636
4-Amino-N-(2-methylphenyl)benzamide
4-Amino-N-(2-methylphenyl)benzamide

Hazard: Irritant

Formula: C₁₄H₁₄N₂O
MDL Number: MFCD00443937
CAS Number: 888-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR