Benzoyl-DL-valine

Supplier: Matrix Scientific

Synonyms: 2-Benzamido-3-methylbutanoic acid

061266-500MG
101921-632EA 243.08 USD
101921-632
Benzoyl-DL-valine
Benzoyl-DL-valine

CAS: 2901-80-6
MDL Number: MFCD00066064
Molecular Formula: C12H15NO3
Molecular Weight: 221,26
Hazard: Irritant

Formula: C₁₂H₁₅NO₃
MW: 221.26 g/mol
MDL Number: MFCD00038283
CAS Number: 2901-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR