2,5-Dimethylbenzophenone

Supplier: Matrix Scientific

061260-500MG
101921-620EA 304.75 USD
101921-620
2,5-Dimethylbenzophenone
2,5-Dimethylbenzophenone

Hazard: Irritant

Formula: C₁₅H₁₄O
CAS Number: 4044-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR