2,3-Dichloromaleic anhydride ≥97%
Supplier: Matrix Scientific
061245-25G
061245-100G
101921-592EA
454.25
USD
101921-592
101921-590
2,3-Dichloromaleic anhydride ≥97%
2,3-Dichloromaleic anhydride
Hazard: Irritant
Formula:
C₄Cl₂O₃ MW: 166.9500 g/mol |
MDL Number:
MFCD00005520 CAS Number: 1122-17-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...