5-Chloro-1-methyl-4-nitroimidazole

Supplier: Matrix Scientific

061234-500MG
101921-568EA 173.65 USD
101921-568
5-Chloro-1-methyl-4-nitroimidazole
5-Chloro-1-methyl-4-nitroimidazole

Hazard: Irritant

Formula: C₄H₄ClN₃O₂
MW: 161.55 g/mol
MDL Number: MFCD00233664
CAS Number: 4897-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR