4-Pyridin-4-yl-1,2,5-oxadiazol-3-amine

Supplier: Matrix Scientific

061229-500MG
101921-558EA 606.15 USD
101921-558
4-Pyridin-4-yl-1,2,5-oxadiazol-3-amine
4-Pyridin-4-yl-1,2,5-oxadiazol-3-amine

Hazard: Irritant

Formula: C₇H₆N₄O
MDL Number: MFCD09701625
CAS Number: 1018143-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR