4-(3-Methoxyphenoxy)-1,2,5-oxadiazol-3-amine

Supplier: Matrix Scientific

061224-500MG
101921-548EA 606.15 USD
101921-548
4-(3-Methoxyphenoxy)-1,2,5-oxadiazol-3-amine
4-(3-Methoxyphenoxy)-1,2,5-oxadiazol-3-amine

Hazard: Irritant

Formula: C₉H₉N₃O₃
MDL Number: MFCD08848253
CAS Number: 924871-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR