(1Z,2E)-N'-Hydroxy-2-(hydroxyimino)pentanimidamide
Supplier: Matrix Scientific
061221-500MG
101921-542EA
606.15
USD
101921-542
(1Z,2E)-N'-Hydroxy-2-(hydroxyimino)pentanimidamide
(1Z,2E)-N'-Hydroxy-2-(hydroxyimino)pentanimidamide
Hazard: Irritant
Formula:
C₅H₁₁N₃O₂ |
MDL Number:
MFCD12197700 CAS Number: 1160264-03-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...