(1Z,2E)-N'-Hydroxy-2-(hydroxyimino)propanimidamide

Supplier: Matrix Scientific

061220-500MG
101921-540EA 606.15 USD
101921-540
(1Z,2E)-N'-Hydroxy-2-(hydroxyimino)propanimidamide
(1Z,2E)-N'-Hydroxy-2-(hydroxyimino)propanimidamide

Hazard: Irritant

Formula: C₃H₇N₃O₂
MDL Number: MFCD12197699
CAS Number: 4937-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR