3-Phenyl-4-[(4-phenyl-1,2,5-oxadiazol-3-yl)-NNO-azoxy]-1,2,5-oxadiazole

Supplier: Matrix Scientific

061218-500MG
101921-536EA 364.62 USD
101921-536
3-Phenyl-4-[(4-phenyl-1,2,5-oxadiazol-3-yl)-NNO-azoxy]-1,2,5-oxadiazole
3-Phenyl-4-[(4-phenyl-1,2,5-oxadiazol-3-yl)-NNO-azoxy]-1,2,5-oxadiazole

Hazard: Irritant

Formula: C₁₆H₁₀N₆O₃
MDL Number: MFCD00603518
CAS Number: 129282-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR