5-Chloro-2,1,3-benzothiadiazole-4-sulfonyl chloride

Supplier: Matrix Scientific

061217-500MG
101921-534EA 484.62 USD
101921-534
5-Chloro-2,1,3-benzothiadiazole-4-sulfonyl chloride
5-Chloro-2,1,3-benzothiadiazole-4-sulfonyl chloride

Hazard: Irritant

Formula: C₆H₂Cl₂N₂O₂S₂
MDL Number: MFCD09028339
CAS Number: 100130-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR