5-Methyl-2,1,3-benzoxadiazole-4-sulfonyl chloride

Supplier: Matrix Scientific

061216-500MG
101921-532EA 484.62 USD
101921-532
5-Methyl-2,1,3-benzoxadiazole-4-sulfonyl chloride
5-Methyl-2,1,3-benzoxadiazole-4-sulfonyl chloride

Hazard: Irritant

Formula: C₇H₅ClN₂O₃S
MDL Number: MFCD11854555
CAS Number: 1152431-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR