2-Amino-N-methyl-N-phenylbenzamide

Supplier: Matrix Scientific

061212-500MG
101921-524EA 183.08 USD
101921-524
2-Amino-N-methyl-N-phenylbenzamide
2-Amino-N-methyl-N-phenylbenzamide

Hazard: Irritant

Formula: C₁₄H₁₄N₂O
MW: 226.2738 g/mol
MDL Number: MFCD00443942
CAS Number: 6632-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR