2-[(2-Chlorobenzyl)sulfonyl]benzoic acid

Supplier: Matrix Scientific

061197-500MG
101921-494EA 243.08 USD
101921-494
2-[(2-Chlorobenzyl)sulfonyl]benzoic acid
2-[(2-Chlorobenzyl)sulfonyl]benzoic acid

Hazard: Irritant

Formula: C₁₄H₁₁ClO₄S
MDL Number: MFCD09905255
CAS Number: 951911-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR