Methyl-4-aminooxanthrene-2-carboxylate

Supplier: Matrix Scientific

061192-500MG
101921-484EA 484.62 USD
101921-484
Methyl-4-aminooxanthrene-2-carboxylate
Methyl-4-aminooxanthrene-2-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₁NO₄
MDL Number: MFCD12033645
CAS Number: 1020242-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR