7-Amino-4,5,6-trimethoxy-2-benzofuran-1(3H)-one

Supplier: Matrix Scientific

061191-500MG
101921-482EA 484.62 USD
101921-482
7-Amino-4,5,6-trimethoxy-2-benzofuran-1(3H)-one
7-Amino-4,5,6-trimethoxy-2-benzofuran-1(3H)-one

Hazard: Irritant

Formula: C₁₁H₁₃NO₅
MDL Number: MFCD10038761
CAS Number: 99856-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR