5-Amino-2,3-dimethyl-2,3-dihydrophthalazine-1,4-dione

Supplier: Matrix Scientific

061186-500MG
101921-472EA 484.62 USD
101921-472
5-Amino-2,3-dimethyl-2,3-dihydrophthalazine-1,4-dione
5-Amino-2,3-dimethyl-2,3-dihydrophthalazine-1,4-dione

Hazard: Irritant

Formula: C₁₀H₁₁N₃O₂
MDL Number: MFCD09064011
CAS Number: 873967-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR