[2-(1-Adamantyl)-4,5-dimethoxyphenyl]amine

Supplier: Matrix Scientific

061183-500MG
101921-466EA 364.62 USD
101921-466
[2-(1-Adamantyl)-4,5-dimethoxyphenyl]amine
[2-(1-Adamantyl)-4,5-dimethoxyphenyl]amine

Hazard: Irritant

Formula: C₁₈H₂₅NO₂
MDL Number: MFCD09787965
CAS Number: 931587-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR