7-(Trifluoromethyl)-3,4-dihydroquinoxalin-2(1H)-one

Supplier: Matrix Scientific

061179-500MG
101921-458EA 484.62 USD
101921-458
7-(Trifluoromethyl)-3,4-dihydroquinoxalin-2(1H)-one
7-(Trifluoromethyl)-3,4-dihydroquinoxalin-2(1H)-one

Hazard: Irritant

Formula: C₉H₇F₃N₂O
MDL Number: MFCD01435942
CAS Number: 716-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR