Ethyl-2-aminoquinoline-3-carboxylate

Supplier: Matrix Scientific

061175-500MG
101921-450EA 484.62 USD
101921-450
Ethyl-2-aminoquinoline-3-carboxylate
Ethyl-2-aminoquinoline-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₂
MDL Number: MFCD01110555
CAS Number: 36926-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR