3-(1-Adamantyl)-4-methoxybenzoic acid

Supplier: Matrix Scientific

061173-500MG
101921-446EA 364.62 USD
101921-446
3-(1-Adamantyl)-4-methoxybenzoic acid
3-(1-Adamantyl)-4-methoxybenzoic acid

Hazard: Irritant

Formula: C₁₈H₂₂O₃
MDL Number: MFCD06014128
CAS Number: 104224-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR