(4-Aminophenyl)(1-methyl-1H-imidazol-2-yl)methanone

Supplier: Matrix Scientific

061170-500MG
101921-440EA 484.62 USD
101921-440
(4-Aminophenyl)(1-methyl-1H-imidazol-2-yl)methanone
(4-Aminophenyl)(1-methyl-1H-imidazol-2-yl)methanone

Hazard: Irritant

Formula: C₁₁H₁₁N₃O
MDL Number: MFCD06655318
CAS Number: 110698-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR