4-(2-Aminoethoxy)-3-methoxybenzoic acid

Supplier: Matrix Scientific

061169-500MG
101921-438EA 349.6 USD
101921-438
4-(2-Aminoethoxy)-3-methoxybenzoic acid
4-(2-Aminoethoxy)-3-methoxybenzoic acid

Hazard: Irritant

Formula: C₁₀H₁₃NO₄
MDL Number: MFCD10039564
CAS Number: 1011408-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR