2-(2-Cyanophenoxy)acetohydrazide

Supplier: Matrix Scientific

061163-500MG
101921-426EA 364.62 USD
101921-426
2-(2-Cyanophenoxy)acetohydrazide
2-(2-Cyanophenoxy)acetohydrazide

Hazard: Irritant

Formula: C₉H₉N₃O₂
MDL Number: MFCD00994328
CAS Number: 347912-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR