2'-Amino-4'-ethoxyacetanilide

Supplier: Matrix Scientific

061161-500MG
101921-422EA 183.08 USD
101921-422
2'-Amino-4'-ethoxyacetanilide
2'-Amino-4'-ethoxyacetanilide

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₂
MDL Number: MFCD00629011
CAS Number: 67169-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR