1-(4-Chlorophenyl)cyclopentanecarboxylic acid

Supplier: Matrix Scientific

Synonyms: 1-(4-Chlorophenyl)-1-cyclopentanecarboxylic acid

061157-500MG
101921-414EA 146.15 USD
101921-414
1-(4-Chlorophenyl)cyclopentanecarboxylic acid
1-(4-Chlorophenyl)cyclopentanecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₃ClO₂
MW: 224.6834 g/mol
MDL Number: MFCD00001373
CAS Number: 80789-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR